Skip to Content
Merck
Hygromycin B from Streptomyces hygroscopicus suitable for plant cell culture, BioReagent, ≥60% (HPLC), lyophilized powder

Hygromycin B from Streptomyces hygroscopicus

Hygromycin B from Streptomyces hygroscopicus suitable for plant cell culture, BioReagent, ≥60% (HPLC), lyophilized powder
Empirical Formula (Hill Notation):
C20H37N3O13
CAS Number:
Molecular Weight:
527.52
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Activity Spectrum
Quality Segment
Application
Mode of Action
Technique