Skip to Content
Merck
Hydroxystilbamidine bis(methanesulfonate) suitable for fluorescence, ≥96.0% (HPLC)

Hydroxystilbamidine bis(methanesulfonate)

Hydroxystilbamidine bis(methanesulfonate) suitable for fluorescence, ≥96.0% (HPLC)
Synonym(s):
2-Hydroxystilbene-4,4′-dicarboxamidine bis(methanesulfonate)
Empirical Formula (Hill Notation):
C18H24N4O7S2
CAS Number:
Molecular Weight:
472.54

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1