
Guanidinium thiocyanate

Synonym(s):
Guanidinium thiocyanate
Linear Formula:
H2NC(NH)NH2 · HSCN
CAS Number:
Molecular Weight:
118.16
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing