
Glycyrrhizin

Synonym(s):
Glycyrrhizic acid, 3-O-(2-O-β-D-Glucopyranuronosyl-α-D-glucopyranuronosyl)-18β-glycyrrhetinic acid, Glycyrrhetinic acid glycoside, Glycyrrhizinic acid
Empirical Formula (Hill Notation):
C42H62O16
CAS Number:
Molecular Weight:
822.93
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing