![Gliclazide impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/182/470/2d2ae51e-ed00-4c0e-978d-f78aade1041e/640/2d2ae51e-ed00-4c0e-978d-f78aade1041e.png)
Gliclazide impurity B
![Gliclazide impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/182/470/2d2ae51e-ed00-4c0e-978d-f78aade1041e/640/2d2ae51e-ed00-4c0e-978d-f78aade1041e.png)
Synonym(s):
2-Nitroso-octahydrocyclopenta[c]pyrrole
Empirical Formula (Hill Notation):
C7H12N2O
CAS Number:
Molecular Weight:
140.18
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing