Skip to Content
Merck
vibrant-m

Gemfibrozil Related Compound A

vibrant-m
Synonym(s):
(EZ)-2,2-Dimethyl-5-[2,5-dimethyl-4-(propene-1-yl)phenoxy]valeric acid
Empirical Formula (Hill Notation):
C18H26O3
CAS Number:
Molecular Weight:
290.40

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1