Fluorouracil Related Compound F
Synonym(s):
5-Fluoro-2-ethoxy-4(1H)pyrimidinone, 2-Ethoxy-5-fluoropyrimidin-4(1H)-one
Linear Formula:
C4N2H2FOOCH2CH3
CAS Number:
Molecular Weight:
158.13
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2