Skip to Content
IL
EN
Back
dTTP
Synonym(s):
Thymidine 5′-triphosphate sodium salt, TTP, dTTP
Empirical Formula (Hill Notation):
C
10
H
16
N
2
O
14
P
3
Na
CAS Number:
18423-43-3
Molecular Weight:
504.15
Products
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
11051482001
dTTP,
Lithium salt
Expand
View Pricing