Skip to Content
Merck
D-(−)-Ribose ≥98%

D-(−)-Ribose

D-(−)-Ribose ≥98%
Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing