Skip to Content
Merck
(+)-O,O′-Di-pivaloyl-D-tartaric acid ≥98.0%

(+)-O,O′-Di-pivaloyl-D-tartaric acid

(+)-O,O′-Di-pivaloyl-D-tartaric acid ≥98.0%
Synonym(s):
(+)-DPTA
Linear Formula:
[(CH3)3CCO2CH(CO2H)-]2
CAS Number:
Molecular Weight:
318.32

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing