![(±)-Nornicotine ≥98% (TLC), liquid](/deepweb/assets/sigmaaldrich/product/structures/382/128/dd86d796-1e7c-4dc5-9784-ea385c5328ff/640/dd86d796-1e7c-4dc5-9784-ea385c5328ff.png)
(±)-Nornicotine
![(±)-Nornicotine ≥98% (TLC), liquid](/deepweb/assets/sigmaaldrich/product/structures/382/128/dd86d796-1e7c-4dc5-9784-ea385c5328ff/640/dd86d796-1e7c-4dc5-9784-ea385c5328ff.png)
Synonym(s):
2-(3-Pyridyl)pyrrolidine, 3-(2-Pyrrolidinyl)pyridine
Empirical Formula (Hill Notation):
C9H12N2
CAS Number:
Molecular Weight:
148.20
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing