Skip to Content
Merck
Dihydrofolic acid ≥90%

Dihydrofolic acid

Dihydrofolic acid ≥90%
Synonym(s):
7,8-Dihydropteroyl-L-glutamic acid, Dihydropteroyl-L-glutamic acid, FAH2
Empirical Formula (Hill Notation):
C19H21N7O6
CAS Number:
Molecular Weight:
443.41

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing