D(+)-2-Phosphoglyceric acid sodium salt hydrate
Synonym(s):
D-Glycerate 2-phosphate sodium salt, Sodium D-2-phosphoglycerate hydrate
Empirical Formula (Hill Notation):
C3H7O7P · xNa+ · yH2O
CAS Number:
Molecular Weight:
186.06 (anhydrous free acid basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing