Dalfampridine Related Compound B
Synonym(s):
4-Amino-3,5-dibromopyridine, 3,5-Dibromo-4-pyridinamine
Empirical Formula (Hill Notation):
C5H4Br2N2
CAS Number:
Molecular Weight:
251.91
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing