
Cyanidin 3,5-diglucoside chloride

Synonym(s):
Cyanin chloride, Cyanidin-3,5-di-O-glucoside
Empirical Formula (Hill Notation):
C27H31ClO16
CAS Number:
Molecular Weight:
646.98
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing