Skip to Content
Merck
Cobalt(II) fluoride tetrahydrate 99.9% trace metals basis

Cobalt(II) fluoride tetrahydrate

Cobalt(II) fluoride tetrahydrate 99.9% trace metals basis
Synonym(s):
Cobalt difluoride hydrate, Cobaltous fluoride tetrahydrate
Linear Formula:
CoF2 · 4H2O
CAS Number:
Molecular Weight:
168.99

Products

Brand
Color
Feature
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing