Clemastine fumarate salt
Synonym(s):
(+)-2-[2-[(p-Chloro-α-methyl-α-phenylbenzyl)oxy]ethyl]-1-methylpyrrolidine fumarate salt, (2R)-2-[2-[(1R)-1-(4-Chlorophenyl)-1-phenylethoxy]ethyl]-1-methylpyrrolidine fumarate salt, 1-Methyl-2-[2-(α-methyl-p-chlorobenzhydryloxy)ethyl]pyrrolidine fumarate salt, 1-Methyl-2-[2-(methyl-p-chlorodiphenylmethyloxy)ethyl]pyrrolidine fumarate salt, Meclastine fumarate salt, Mecloprodine
Empirical Formula (Hill Notation):
C21H26ClNO · C4H4O4
CAS Number:
Molecular Weight:
459.96
Products
Product Category
Brand
Color
Feature
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing