
Cirsimarin

Synonym(s):
4′,5-Dihydroxy-6,7-dimethoxyflavone 4′-glucoside, Cirsimaretin, Cirsimaritin 4′-glucoside, Cirsitakaoside
Empirical Formula (Hill Notation):
C23H24O11
CAS Number:
Molecular Weight:
476.43
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing