![Ceftiofur VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/200/530/e53f8b63-754c-44ed-97bb-bff2f7e783ca/640/e53f8b63-754c-44ed-97bb-bff2f7e783ca.png)
Ceftiofur
![Ceftiofur VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/200/530/e53f8b63-754c-44ed-97bb-bff2f7e783ca/640/e53f8b63-754c-44ed-97bb-bff2f7e783ca.png)
Synonym(s):
3-[(2-Furylcarbonyl)thio]methyl]-7-[2-(2-amino-4-thiazolyl)-2-(methoxyiminoacetamido)-3-cephem-4-carboxylic acid
Empirical Formula (Hill Notation):
C19H17N5O7S3
CAS Number:
Molecular Weight:
523.56
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing