Skip to Content
Merck
vibrant-m

Cathinone hydrochloride

vibrant-m
Empirical Formula (Hill Notation):
C9H11NO · HCl
CAS Number:
Molecular Weight:
185.65

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing