Skip to Content
Merck
Butyl p-hydroxybenzoate phyproof® Reference Substance

Butyl p-hydroxybenzoate

Butyl p-hydroxybenzoate phyproof® Reference Substance
Synonym(s):
Butylparaben, 4-Hydroxybenzoic acid butyl ester
Empirical Formula (Hill Notation):
C11H14O3
CAS Number:
Molecular Weight:
194.23

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing