Skip to Content
Merck
Boc-Pip-OH 98%

Boc-Pip-OH

Boc-Pip-OH 98%
Synonym(s):
(S)-(−)-1-(tert-Butoxycarbonyl)-2-piperidinecarboxylic acid, (S)-1-Boc-piperidine-2-carboxylic acid, N-Boc-L-pipecolinic acid
Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing