ATM Kinase Inhibitor
Synonym(s):
KU-55933, 2-(4-Morpholinyl)-6-(1-thianthrenyl)-4H-Pyran-4-one, 2-(Morpholin-4-yl)-6-(thianthren-1-yl)-4H-pyran-4-one
Empirical Formula (Hill Notation):
C21H17NO3S2
CAS Number:
Molecular Weight:
395.49
Products
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing