Skip to Content
Merck
(−)-Asarinin ≥98% (HPLC)

(−)-Asarinin

(−)-Asarinin ≥98% (HPLC)
Synonym(s):
(-)-Episesamin
Empirical Formula (Hill Notation):
C20H18O6
CAS Number:
Molecular Weight:
354.35

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing