![Arjungenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/383/178/a3a1c158-1193-463a-ae49-e8bbe6da18e5/640/a3a1c158-1193-463a-ae49-e8bbe6da18e5.png)
Arjungenin
![Arjungenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/383/178/a3a1c158-1193-463a-ae49-e8bbe6da18e5/640/a3a1c158-1193-463a-ae49-e8bbe6da18e5.png)
Synonym(s):
(2α,3β,49α)- 2,3,19,23-Tetrahydroxyolean-12-en-28-oic acid, 2α,19α,23-Trihydroxyoleanolic acid, Arjugenin
Empirical Formula (Hill Notation):
C30H48O6
CAS Number:
Molecular Weight:
504.70
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing