Anti-WDR13 antibody produced in rabbit
Synonym(s):
Anti-WD repeat domain 13, Anti-WD repeat protein 13 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique