ANTI-PNP antibody produced in rabbit
Synonym(s):
Anti-Inosine phosphorylase, Anti-NP, Anti-PNP, Anti-Purine nucleoside phosphorylase
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1