![Anti-phospho-ATF-2 (pThr69,71) antibody, Mouse monoclonal clone ATF-22P, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/201/944/848ff16e-e6df-4ec1-8e3d-93d11fe4218a/640/848ff16e-e6df-4ec1-8e3d-93d11fe4218a.jpg)
Anti-phospho-ATF-2 (pThr69,71) antibody, Mouse monoclonal
![Anti-phospho-ATF-2 (pThr69,71) antibody, Mouse monoclonal clone ATF-22P, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/201/944/848ff16e-e6df-4ec1-8e3d-93d11fe4218a/640/848ff16e-e6df-4ec1-8e3d-93d11fe4218a.jpg)
Synonym(s):
Monoclonal Anti-phospho-ATF-2 (pThr69,71)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing