Skip to Content
Merck
Antipain, Dihydrochloride Peptidyl arginine aldehyde protease inhibitor produced by actinomycetes.

Antipain dihydrochloride

Antipain, Dihydrochloride Peptidyl arginine aldehyde protease inhibitor produced by actinomycetes.
Synonym(s):
N-(Nα-Carbonyl-Arg-Val-Arg-al)-Phe, Antipain dihydrochloride from microbial source, Antipain, Dihydrochloride
Empirical Formula (Hill Notation):
C27H44N10O6 · 2HCl
CAS Number:
Molecular Weight:
677.62

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Research Area
Quality Segment
Mode of Action
Technique