![Anti-NCAPH antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/194/497/b72dae5e-9ce5-441c-8774-80c0dc5ed745/640/b72dae5e-9ce5-441c-8774-80c0dc5ed745.jpg)
Anti-NCAPH antibody produced in rabbit
![Anti-NCAPH antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/194/497/b72dae5e-9ce5-441c-8774-80c0dc5ed745/640/b72dae5e-9ce5-441c-8774-80c0dc5ed745.jpg)
Synonym(s):
Anti-Barren homolog protein 1 antibody produced in rabbit, Anti-Chromosome-associated protein H antibody produced in rabbit, Anti-Condensin complex subunit 2 antibody produced in rabbit, Anti-Non-SMC condensin I complex subunit H antibody produced in rabbit, Anti-XCAP-H homolog antibody produced in rabbit, Anti-hCAP-H antibody produced in rabbit, Anti-BRRN1, Anti-CAP-H, Anti-HCAP-H, Anti-Non-SMC condensin I complex, subunit H, Barren homolog protein 1, Chromosome-associated protein H, Condensin complex subunit 2, Non-SMC condensin I complex subunit H, hCAP-H
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing