![Anti-CD2AP antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/385/989/ac1c7f03-5376-4f51-84b0-90cfc6e50338/640/ac1c7f03-5376-4f51-84b0-90cfc6e50338.jpg)
Anti-CD2AP antibody produced in rabbit
![Anti-CD2AP antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/385/989/ac1c7f03-5376-4f51-84b0-90cfc6e50338/640/ac1c7f03-5376-4f51-84b0-90cfc6e50338.jpg)
Synonym(s):
Anti-Adapter protein CMS antibody produced in rabbit, Anti-CD2-associated protein antibody produced in rabbit, Anti-Cas ligand with multiple SH3 domains antibody produced in rabbit, CD2AP, CMS, DKFZp586H0519
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing