Skip to Content
Merck
Anthracene sublimed grade, ≥99%

Anthracene

Anthracene sublimed grade, ≥99%
Synonym(s):
Anthraxcene, Paranaphthalene, Anthracene
Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique