Skip to Content
Merck
Alverine citrate salt European Pharmacopoeia (EP) Reference Standard

Alverine citrate salt

Alverine citrate salt European Pharmacopoeia (EP) Reference Standard
Synonym(s):
N-Ethyl-3,3′-diphenyldipropylamine, NSC 35459
Empirical Formula (Hill Notation):
C20H27N · C6H8O7
CAS Number:
Molecular Weight:
473.56

Products

Product Category
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing