Skip to Content
Merck
Alprazolam solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Alprazolam solution

Alprazolam solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C17H13ClN4
CAS Number:
Molecular Weight:
308.76
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1