![α-D-Glucose 1-phosphate dipotassium salt hydrate ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/415/358/bffa04f4-4417-4431-8949-6d4937a19f10/640/bffa04f4-4417-4431-8949-6d4937a19f10.png)
α-D-Glucose 1-phosphate dipotassium salt hydrate
![α-D-Glucose 1-phosphate dipotassium salt hydrate ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/415/358/bffa04f4-4417-4431-8949-6d4937a19f10/640/bffa04f4-4417-4431-8949-6d4937a19f10.png)
Synonym(s):
α-D-Glucopyranose 1-phosphate, Cori ester
Empirical Formula (Hill Notation):
C6H11K2O9P · xH2O
CAS Number:
Molecular Weight:
336.32 (anhydrous basis)
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing