![Abacavir Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/125/112/5ee54458-cac0-4239-9ad6-5bda1ee03e64/640/5ee54458-cac0-4239-9ad6-5bda1ee03e64.jpg)
Abacavir Related Compound A
![Abacavir Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/125/112/5ee54458-cac0-4239-9ad6-5bda1ee03e64/640/5ee54458-cac0-4239-9ad6-5bda1ee03e64.jpg)
Synonym(s):
(1S,4R)-4-[2,6-Diamino-9H-purin-9-yl]-2-cyclopentene-1-methanol
Empirical Formula (Hill Notation):
C11H14N6O
CAS Number:
Molecular Weight:
246.27
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2