7-Allyl-7,8-dihydro-8-oxoguanosine
Empirical Formula (Hill Notation):
C13H17N5O6
CAS Number:
Molecular Weight:
339.30
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing