5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt
Synonym(s):
X-GlcA, X-glucuronide CHA salt
Empirical Formula (Hill Notation):
C14H13BrClNO7 · C6H13N
CAS Number:
Molecular Weight:
521.79
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing