Skip to Content
Merck
vibrant-m

5′-Adenylic acid

vibrant-m
Synonym(s):
Adenosine 5′-monophosphate, 5′-Adenylic Acid, AMP
Empirical Formula (Hill Notation):
C10H14N5O7P
CAS Number:
Molecular Weight:
347.22

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing