Skip to Content
Merck
4-Iodo-L-phenylalanine

4-Iodo-L-phenylalanine

4-Iodo-L-phenylalanine
Synonym(s):
(S)-2-Amino-3-(4-iodophenyl)propanoic acid
Empirical Formula (Hill Notation):
C9H10INO2
CAS Number:
Molecular Weight:
291.09

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing