Skip to Content
Merck
4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one analytical standard, for drug analysis

4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one

4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one analytical standard, for drug analysis
Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique