Skip to Content
Merck
3-(Trifluoromethyl)benzenesulfonyl chloride

3-(Trifluoromethyl)benzenesulfonyl chloride

3-(Trifluoromethyl)benzenesulfonyl chloride
Linear Formula:
CF3C6H4SO2Cl
CAS Number:
Molecular Weight:
244.62

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing