Skip to Content
Merck
3-Mercapto-1-hexanol ≥95%

3-Mercapto-1-hexanol

3-Mercapto-1-hexanol ≥95%
Empirical Formula (Hill Notation):
C6H14OS
CAS Number:
Molecular Weight:
134.24

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing