Skip to Content
Merck
2-Phenylpropyl isobutyrate ≥97%, FG

2-Phenylpropyl isobutyrate

2-Phenylpropyl isobutyrate ≥97%, FG
Synonym(s):
Hydratropyl isobutyrate
Linear Formula:
(CH3)2CHCO2CH2CH(C6H5)CH3
CAS Number:
Molecular Weight:
206.28
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing