Skip to Content
Merck
2-Ketoglutaric acid-1-13C API for Clinical Studies, ≥99 atom % 13C

2-Ketoglutaric acid-1-13C

2-Ketoglutaric acid-1-13C API for Clinical Studies, ≥99 atom % 13C
Synonym(s):
α-Ketoglutaric acid-1-13C, 2-Ketopentanedioic acid-1-13C, 2-Oxoglutaric acid-1-13C, 2-Oxopentanedioic acid-1-13C
Linear Formula:
HOCOCH2CH2CO13CO2H
CAS Number:
Molecular Weight:
147.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2