
2-Fluoro-DL-α-phenylglycine

Linear Formula:
FC6H4CH(NH2)COOH
CAS Number:
Molecular Weight:
169.15
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing