![1-Naphthylamine analytical standard](/deepweb/assets/sigmaaldrich/product/structures/186/377/0d9b21d5-dcc8-42f7-ae3b-187befc4b7a6/640/0d9b21d5-dcc8-42f7-ae3b-187befc4b7a6.png)
1-Naphthylamine
![1-Naphthylamine analytical standard](/deepweb/assets/sigmaaldrich/product/structures/186/377/0d9b21d5-dcc8-42f7-ae3b-187befc4b7a6/640/0d9b21d5-dcc8-42f7-ae3b-187befc4b7a6.png)
Synonym(s):
α-Naphthylamine, 1-Aminonaphthalene, 1-Naphthylamine
Linear Formula:
C10H7NH2
CAS Number:
Molecular Weight:
143.19
Products
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing