Skip to Content
Merck
1,3-Bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene

1,3-Bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene

1,3-Bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-2-ylidene
Synonym(s):
1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene, SIMes
Empirical Formula (Hill Notation):
C21H26N2
CAS Number:
Molecular Weight:
306.44

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing