Skip to Content
Merck
All Photos(3)

Documents

V900309

Sigma-Aldrich

MOPS hemisodium salt

Vetec, reagent grade, 98%

Synonym(s):

3-(N-Morpholino)propanesulfonic acid hemisodium salt, 4-Morpholinepropanesulfonic acid hemisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14.5NO4S · 0.5Na
CAS Number:
Molecular Weight:
220.25
UNSPSC Code:
12161700

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

useful pH range

6.5-7.9

pKa (25 °C)

7.2

solubility

water: 0.5 g/mL, clear, colorless to faintly yellow

SMILES string

[Na+].OS(=O)(=O)CCCN1CCOCC1.[O-]S(=O)(=O)CCCN2CCOCC2

InChI

1S/2C7H15NO4S.Na/c2*9-13(10,11)7-1-2-8-3-5-12-6-4-8;/h2*1-7H2,(H,9,10,11);/q;;+1/p-1

InChI key

OVCWZKQPQOVIDT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service