Key Documents
20839-U
Supelco
Needle Guide
About This Item
UNSPSC Code:
41115717
packaging
pkg of 1 ea
General description
Needle guide in the septum cap directs the syringe needle to pierce the septum at the same spot for every injection. Autosamplers are provided with these to hold amd support the needle.
Prolongs septum and needle life. Use with septum nuts that have a 3/16 in. diameter hole in the center. For septum nuts with smaller holes, simply drill to 3/16 in. to insert guide.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Konrad Grob
Split and Splitless Injection for Quantitative Gas Chromatography, 426-426 (2008)